Tisk…

Anti-MC4 Receptor Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3936-100EA 6910 CZK
BIOV3936-100
Anti-MC4 Receptor Rabbit Polyclonal Antibody
Protilátky
MC4R (Melanocortin-4 receptor) belongs to the GPCR superfamily that is responsible for multiple signal transduction pathway which includes the cAMP and MAPK signaling pathways. Expression of MC4R has been reported in brain. From genetic studies of mice and humans, it was established that MC4 receptor plays a critical role in appetite regulation. Mutation in the MC4R gene is associated with obesity in humans.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MC4 Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR