Tisk…

Anti-Beta3-AR Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3939-200EA 7900 CZK
BIOV3939-200
Anti-Beta3-AR Rabbit Polyclonal Antibody
Protilátky
The neurotransmitter/hormone adrenaline (epinephrine, adrenalin) plays a central role in the mammalian stress response, increasing heart rate, raising blood pressure, and increasing blood glucose levels. Adrenaline activates both alpha-adrenergic receptors and beta-adrenergic receptors. There are mainly three subtypes of beta adrenergic receptors known, beta1, beta2, and beta3. These receptors are expressed primarily in the heart, respiratory tissue, and adipose tissue. Beta3-receptors are abundant in brown adipocytes and play vital roles in lipolysis and thermoregulation. Beta adrenergic receptors are gaining much interest among researches in the studies of type-2 diabetes mellitus and obesity. It is also being considered as a therapeutic target for heart failure.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Beta3-AR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR