Tisk…

Anti-AdipoR1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3940-200EA 7330 CZK
BIOV3940-200
Anti-AdipoR1 Rabbit Polyclonal Antibody
Protilátky
The adiponectin receptors, ADIPOR1 and ADIPOR2 are receptors for globular and full-length adiponectin. These receptors mediate the increase of AMPK and PPAR-α ligand activities, as well as fatty acid oxidation and glucose uptake by adiponectin.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: AdipoR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR