Tisk…

Anti-C/EBP alpha Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3943-200EA 6910 CZK
BIOV3943-200
Anti-C/EBP alpha Rabbit Polyclonal Antibody
Protilátky
C/EBP alpha helps regulate terminal adipocyte differentiation by turning on fat-specific genes required for the synthesis, uptake, and storage of long chain fatty acids. Studies have shown that C/EBP delta and C/EBP beta play an early catalytic role in the differentiation pathway, relaying the effects of the hormonal stimulants DEX and MIX in a cascade-like fashion, leading to the activation of the gene encoding C/EBP alpha.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: C/EBP alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR