Tisk…

Anti-Pyruvate Dehydrogenase Phosphatase 2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3944-200EA 6910 CZK
BIOV3944-200
Anti-Pyruvate Dehydrogenase Phosphatase 2 Rabbit Polyclonal Antibody
Protilátky
PDP2, Pyruvate dehydrogenase phosphatase 2, catalyzes the dephosphorylation and activation of E1 component to reverse the effects of pyruvate dehydrogenase kinases. Pyruvate dehydrogenase (E1) is one of the three components (E1, E2, and E3) of the large pyruvate dehydrogenase complex. 

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Pyruvate Dehydrogenase Phosphatase 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR