Tisk…
Anti-FTO Rabbit Polyclonal Antibody
Dodavatel: BioVision
BIOV6764-100EA 7780 CZK
BIOV6764-100
Anti-FTO Rabbit Polyclonal Antibody
FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse ‘fused toes’ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.
Recommended Dilutions: Western Blot: ~1:1000, IHC: ~1:50-1:100, FACS: ~1:10–1:50.
Type: Primary
Antigen: FTO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human.