Tisk…

Anti-Sox-2 Mouse Monoclonal Antibody [clone: 57CT23.3.4]

Dodavatel: BioVision
BIOV6772-100EA 7470 CZK
BIOV6772-100
Anti-Sox-2 Mouse Monoclonal Antibody [clone: 57CT23.3.4]
Protilátky
Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes clusters at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of this family have been shown to be conserved during evolution and to play key roles during animal development. Some are involved in human diseases, including sex reversal.

Recommended Dilutions: Western Blot: ~1:200-1:2000, IHC: 1:50-1:100, FACS: 1:10-1:50, Immunofluorescence: 1:100.

Type: Primary
Antigen: Sox-2
Clonality: Monoclonal
Clone: 57CT23.3.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human.
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR