Tisk…

Anti-Runx2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6773-100EA 7780 CZK
BIOV6773-100
Anti-Runx2 Rabbit Polyclonal Antibody
The mammalian Runt-related transcription factor (RUNX) family comprises three members, RUNX1 (also designated AML-1, PEBP2åB, CBFA2), RUNX2 (also designated AML-3, PEBP2åA, CBFA1, Osf2) and RUNX3 (also designated AML-2, PEBPåC, CBFA3;RUNX family members are DNA-binding proteins that regulate the expression of genes involved in cellular differentiation and cell cycle progression. RUNX2 is essential for skeletal mineralization in that it stimulates osteoblast differentiation of mesenchymal stem cells, promotes chondrocyte hypertrophy and contributes to endothelial cell migration and vascular invasion of developing bones. Regulating RUNX2 expression may be a useful therapeutic tool for promoting bone formation. Mutations in the C-terminus of RUNX2 are associated with cleidocranial dysplasia syndrome, an autosomal-dominant skeletal dysplasia syndrome that is characterized by widely patent calvarial sutures, clavicular hypoplasia, supernumerary teeth, and short stature.

Recommended Dilutions: Western Blot: ~1:1000, IHC: 1:10-1:50, FACS: 1:10-1:50.

Type: Primary
Antigen: Runx2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human.
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR