Tisk…

Anti-VCAM-1/CD106 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6783-100EA 6910 CZK
BIOV6783-100
Anti-VCAM-1/CD106 Rabbit Polyclonal Antibody
Protilátky
Cell adhesion molecules are a family of closely related cell surface glycoproteins involved in cell-cell interactions during growth and are thought to play an important role in embryogenesis and development. Neuronal cell adhesion molecule (NCAM) expression is observed in a variety of human tumors including neuroblastoma, rhabdomyosarcoma, Wilms’ tumors, Ewing’s sarcomas and some primitive myeloid malignancies. The intracellular adhesion molecule-1 (ICAM-1), also referred to as CD54, is an integral membrane protein of the immunoglobulin superfamily and recognizes the B2å1 and B2åM integrins. PECAM-1 (platelet/endothelial cell adhesion molecule-1), also referred to as CD31, is a glycoprotein expressed on the cell surfaces of monocytes, neutrophils, platelets and a subpopulation of T cells. VCAM-1 (vascular cell adhesion molecule-1) was first identified as an adhesion molecule induced on human endothelial cells by inflammatory cytokines such as IL-1, tumor necrosis factor (TNF) and lipopolysaccharide (LPS;The KALIG gene encodes a nerve cell adhesion molecule (NCAM) -like protein and is deleted in 66% of patients with Kallmann's syndrome, anosmia with secondary hypogonadism.

Recommended Dilutions: Western Blot: 1:1000.

Type: Primary
Antigen: VCAM-1/CD106
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse.
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR