Tisk…

Anti-CD105/Endoglin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6800-100EA 6910 CZK
BIOV6800-100
Anti-CD105/Endoglin Rabbit Polyclonal Antibody
Protilátky
Endoglin (ENG, CD105) is an auxiliary receptor for the TGF-β receptor complex, functioning in related signaling pathways. Endoglin is a transmembrane protein that exists as a disulfide-linked homodimer. It is mainly expressed in vascular and connective tissues and in endothelial and stromal cells. Upregulated endoglin expression has been reported during wound healing and tumor vascularization, and in inflammatory tissues and developing embryos. Mutations in endoglin have been found to be a causal factor of hereditary hemorrhagic telangiectasia (HHT), a disease characterized by malformation of vascular structure. The importance of this protein for normal and tumor vascular function makes it a good marker for endothelial cell proliferation as well as a potential therapeutic target in cancer.

Recommended Dilutions: Western Blot: 1:1000, IHC: 1:10-1:50, FC: 1:10-1:50.

Type: Primary
Antigen: CD105/Endoglin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse.
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR