Tisk…

Anti-AS160/TBC1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3954-100EA 6910 CZK
BIOV3954-100
Anti-AS160/TBC1 Rabbit Polyclonal Antibody
Protilátky
AS160 is a 160 kDa Akt serine/threonine kinase substrate that plays a critical role in regulating insulin-stimulated exocytosis of glucose transporter GLUT4. Increased in insulin treatment stimulate phophorylation of AS160 that regulates GLUT4 translocation via inactivation of the Rab GTPase function.

Recommended Dilutions: Western Blot: 1–4 µg/ml).

Type: Primary
Antigen: AS160/TBC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR