Tisk…

Anti-PI3 kinase Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3959-100EA 7780 CZK
BIOV3959-100
Anti-PI3 kinase Rabbit Polyclonal Antibody
Phosphatidylinositol 3-kinase (PI 3-kinase), is a phosphoinositide-specific protein kinase that plays a central role in mitogenic signal transduction leading to cellular regulation, cell growth and differentiation, and oncogenesis. PI 3-kinase associates with, and is activated by a wide range of tyrosine kinase receptors including EGF, PDGF, CSF-1, erbB-3 and insulin receptor, non-receptor protein-tyrosine kinases of the src family (e.g. pp60src), and crk and abl proto-oncogene products. PI 3-kinase is involved in signal transduction in terminally differentiated, activated cells like platelets and neutrophils.

Recommended Dilutions: Western Blot: 1-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: PI3 kinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR