Tisk…

Anti-MMP-9 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3969-200EA 6910 CZK
BIOV3969-200
Anti-MMP-9 Rabbit Polyclonal Antibody
Protilátky
The matrix metalloproteinases (MMP) are a family of peptidase enzymes responsible for the degradation of extracellular matrix components, including collagen, gelatin, fibronectin, laminin and proteoglycan. Transcription of MMP genes is differentially activated by phorbol ester, lipopolysaccharide (LPS) or staphylococcal enterotoxin B (SEB;MMP catalysis requires both calcium and zinc. MMP-9 has been shown to degrade bone collagens in concert with MMP-1 (also designated interstitial collagenase, fibroblast collagenase or collagenase-1), and cysteine proteases and may play a role in bone osteoclastic resorption.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: MMP-9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR