Tisk…

Anti-SBP-Tag Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3986-100EA 8600 CZK
BIOV3986-100
Anti-SBP-Tag Rabbit Polyclonal Antibody
Protilátky
Streptavidin Binding Peptide (SBP) tag is very versatile and can be inserted into expression vectors for easy detection of recombinant proteins and purification by use of affinity columns. The full sequence for SBP-tag is 38 amino acids long and exhibits a nanomolar scale binding affinity for streptavidin.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SBP-Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR