Tisk…

Anti-C-Myc Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3995-100EA 7190 CZK
BIOV3995-100
Anti-C-Myc Rabbit Polyclonal Antibody
Protilátky
Epitope tags are usually for the labeling and detection of proteins using immunoblotting, immunoprecipitation and immunostaining techniques. Due to their small size, they are unlikely to affect the tagged protein’s biochemical properties. The Myc epitope tag is widely used to detect expression of recombinant proteins in bacteria, yeast, insect and mammalian systems.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunoprecipitation: 20-40 µg/ml; and Immunocytochemistry: 10-20 µg/ml

Type: Primary
Antigen: C-Myc
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR