Tisk…

Anti-6xHis Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3998-100EA 7900 CZK
BIOV3998-100
Anti-6xHis Rabbit Polyclonal Antibody
Protilátky
A variety of plasmids contain DNA that encodes an N-terminal tag consisting of six histidine (His) residues, followed by an extended multiple cloning sites. The 6xHis tag on the expressed recombinant proteins allows for efficient coupling to Ni++ affinity resins and purification by a single step chromatography. As is the case with other protein tags, this polyhistidine tag can be cleaved at sites recognized by enzymes such as thrombin and enterokinases to isolate the protein of interest.

Recommended Dilutions: Western Blot: 1 µg/ml; Immunoprecipitation: 40 µg/ml; and Immunocytochemistry: 2 µg/ml

Type: Primary
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR