Tisk…

Anti-Sushi Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5010-100EA 10350 CZK
BIOV5010-100
Anti-Sushi Rabbit Polyclonal Antibody
Sushi Peptide S3 is a trimer of one of the high endotoxin-binding domains, Sushi 3 (or S3) within Factor C, a lipopolysaccharide (LPS)-sensitive serine protease of the horseshoe crab (Limulus Polyphemus;S3 display detergent-like properties in disrupting LPS aggregates, with specificity for palmitoyl-oleoyl-phosphatidylglycerol (POPG) resulting from electrostatic and hydrophobic forces between the peptides and the bacterial lipids. The unsaturated nature of POPG confers fluidity and enhances insertion of the peptides into the lipid bilayer, causing maximal disruption of the bacterial membrane. In short, peptide S3 can bind to lipopolysaccharide (LPS) and inhibit the growth of Gram-negative bacteria without affecting mammalian cells. It has been shown that endotoxin activates Factor C based catalytic coagulation cascade resulting in the gelation of Limulus blood. This process is the basis of Limulus Amebocyte Lysate (LAL) endotoxin detection method.

Recommended Dilutions: Western Blot: 1-4 µg/ml;

Type: Primary
Antigen: Sushi
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR