Tisk…

Anti-CT-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5011-100EA 6910 CZK
BIOV5011-100
Anti-CT-1 Rabbit Polyclonal Antibody
Protilátky
Human Cardiotrophin-1 (CT-1) is a member of the leukemia inhibitory/ciliary neurotrophic factor/Oncostatin M/interleukin-6/interleukin-11 family of cytokines. Cardiotrophin-1 affects cells throμgh interaction with the gp 130 receptor subunit. Recombinant human Cardiotrophin-1 is a 22.5 kDa protein containing 200 amino acid residues.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: CT-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR