Tisk…

Anti-CNTF Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5017-100EA 6910 CZK
BIOV5017-100
Anti-CNTF Rabbit Polyclonal Antibody
Protilátky
CNTF (Ciliary Neurotrophic Factor) is a potent neural factor that was originally characterized as a survivability factor for chick ciliary neurons in vitro. CNTF has also been shown to promote survivability and differentiation of other neuronal cell types. CNTF is a 22.9 kDa protein containing 200 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CNTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR