Tisk…

Anti-EGF Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5023R-100EA 7780 CZK
BIOV5023R-100
Anti-EGF Rabbit Polyclonal Antibody
EGF (Epidermal Growth Factor) is polypeptide growth factor that stimulates the proliferation of a wide range of epidermal and epithelial cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR