Tisk…

Anti-Eotaxin/CCL11 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5029-100EA 8430 CZK
BIOV5029-100
Anti-Eotaxin/CCL11 Rabbit Polyclonal Antibody
Eotaxin is a 74-amino acid eosinophil-chemotactic CC chemokine originally found in bronchoalveolar lavage fluid from allergic inflammatory subjects. It is involved in regulating the recruitment and activation of inflammatory leukocytes, particularly eosinophils. It may play a fundamental role in the development of allergic responses.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Eotaxin/CCL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR