Tisk…

Anti-FGF-10 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5060A-100EA 6910 CZK
BIOV5060A-100
Anti-FGF-10 Rabbit Polyclonal Antibody
Protilátky
Fibroblast Growth Factor-10 (also called KGF-2) is a heparin binding growth factor that stimulates the proliferation and activation of cells that express FGF receptors. FGF-10 is mostly related to FGF-7/KGF and is expressed during development and preferentially in adult lungs.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: FGF-10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR