Tisk…

Anti-Flt3 Ligand Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5085R-100EA 6910 CZK
BIOV5085R-100
Anti-Flt3 Ligand Rabbit Polyclonal Antibody
Protilátky
Human flt3-Ligand (flk2-ligand) stimulates the proliferation and colony formation of certain bone marrow precursor cells including CD34+ cells. The antibody was produced using the recombinant human flt3-ligand as immunogen, which is a soluble 17.6 kDa polypeptide containing 155 amino acid residues, that comprises the extracellular domain of the transmembrane flt3-ligand protein.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Flt3 Ligand
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR