Tisk…

Anti-IGF-I Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5119-100EA 6910 CZK
BIOV5119-100
Anti-IGF-I Rabbit Polyclonal Antibody
Protilátky
IGF-I (Insulin-like Growth Factor-I) is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types including muscle, bone, and cartilage tissue. Human IGF-I is a 7.6 kDa protein containing 70 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IGF-I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR