Tisk…

Anti-Anti-Rat IGF-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5121-100EA 7780 CZK
BIOV5121-100
Anti-Anti-Rat IGF-1 Rabbit Polyclonal Antibody
IGF-I (insulin-like Growth Factor-I) is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types including muscle, bone, and cartilage tissue. Rat IGF-I is a 7.69 kDa protein.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Anti-Rat IGF-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR