Tisk…
Anti-IL-1 alpha Rabbit Polyclonal Antibody
Dodavatel: BioVision
BIOV5125-100EA 7780 CZK
BIOV5125-100
Anti-IL-1 alpha Rabbit Polyclonal Antibody
IL-1α is a potent immuno-modulator that mediates a wide range of immune and inflammatory responses. Human IL-1α is an 18.0 kDa protein containing 159 amino acid residues.
Recommended Dilutions: Western Blot: 0.5-2 µg/ml
Type: Primary
Antigen: IL-1 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human