Tisk…

Anti-IL-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5131-100EA 7780 CZK
BIOV5131-100
Anti-IL-2 Rabbit Polyclonal Antibody
IL-2 is a potent immuno-modulator, which mediates a wide range of immune and inflammatory responses. Human IL-2 is a 15.5 kDa protein containing 134 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IL-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR