Tisk…

Anti-IL-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5133-100EA 6910 CZK
BIOV5133-100
Anti-IL-2 Rabbit Polyclonal Antibody
Protilátky
Interleukin-2 (IL-2) is a potent lymphoid cell growth factor that exerts its biological activity primarily on T cells. The antibody detects a 17.5 kDa recombinant rat IL-2 protein containing 134 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IL-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR