Tisk…

Anti-IL-6 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5143-100EA 6910 CZK
BIOV5143-100
Anti-IL-6 Rabbit Polyclonal Antibody
Protilátky
IL-6 is a potent lymphoid cell growth factor that stimulates the growth and survivability of certain B cells and T cells. Human IL-6 is a 20.9 kDa protein containing 184 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4.0 µg/ml

Type: Primary
Antigen: IL-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR