Tisk…

Anti-IL-6 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5144-100EA 7470 CZK
BIOV5144-100
Anti-IL-6 Rabbit Polyclonal Antibody
Protilátky
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant murine IL-6. The antibody was purified by affinity chromatography employing immobilized murine IL-6 matrix.

Recommended Dilutions: ELISA: 1-3 µg/ml; Western Blot: 0.5-1 µg/ml; and Neutralization: 0.1-0.5 µg/ml

Type: Primary
Antigen: IL-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR