Tisk…

Anti-IL-7 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5148-100EA 6910 CZK
BIOV5148-100
Anti-IL-7 Rabbit Polyclonal Antibody
Protilátky
IL-7 is a hematopoietic growth factor which affects primarily early B and T cells. Produced by thymic stromal cells, spleen cells and keratinocytes, IL-7 can also co-stimulate the proliferation of mature T cells in combination with other factors such as ConA and IL-2. Human and murine IL-7 is cross-species reactive.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IL-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR