Tisk…

Anti-IL-8 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5149-100EA 7470 CZK
BIOV5149-100
Anti-IL-8 Rabbit Polyclonal Antibody
Protilátky
IL-8 is a proinflammatory CXC chemokine that can signal throμgh the CXCR1 and CXCR2 receptors. It is secreted by monocytes and endothelial cells. IL-8 chemoattracts and activates neutrophils.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml

Type: Primary
Antigen: IL-8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR