Tisk…

Anti-IL-15 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5172-100EA 8430 CZK
BIOV5172-100
Anti-IL-15 Rabbit Polyclonal Antibody
IL-15 is a member of the four alpha-helical bundle family of cytokines with a molecular weight of 14-15 kDa. Its in vitro activity is very similar to IL-2, except that IL-15 utilizes the IL-15 receptor alpha subunit for binding. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes. It also activates a number of important intracellular signaling molecules. This implies that IL-15 could be essential for the immune responses, allograpt rejection, and the pathogenesis of autoimmune diseases.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml

Type: Primary
Antigen: IL-15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR