Tisk…

Anti-IL-18 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5179-100EA 7190 CZK
BIOV5179-100
Anti-IL-18 Rabbit Polyclonal Antibody
Protilátky
Interleukin-18 (IL-18), an 18 kDa cytokine, is a single, non-glycosylated polypeptide chain containing 157 amino acids. IL-18 was previously known as interferon-g inducing factor (IGIF;This pleiotropic cytokine is produced by monocyte/macrophage cells. Like IL-12, IL-18 plays an important role in cell-mediated immune responses.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IL-18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR