Tisk…

Anti-IP-10 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5200-100EA 8740 CZK
BIOV5200-100
Anti-IP-10 Rabbit Polyclonal Antibody
Protilátky
Human IP-10 is an 8.5 kDa protein produced by several cell types during the delayed type hypersensitivity response. IP-10 acts as a chemoattractant towards monocytes, lymphocytes, and certain T cells.

Recommended Dilutions: Western Blot: 0.5-1.0 µg/ml; Neutralization: 10 µg/ml; and ELISA: 2-4 µg/ml)

Type: Primary
Antigen: IP-10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR