Tisk…

Anti-MCP-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5222-100EA 6910 CZK
BIOV5222-100
Anti-MCP-1 Rabbit Polyclonal Antibody
Protilátky
Human macrophage/monocyte chemotactic and activating factor (MCAF), also called human monocyte chemotactic protein-1 (MCP-1), is an 8.6 kDa protein containing 76 amino acid residues. It plays an important role in the inflammatory response of blood monocytes and tissue microphages.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MCP-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR