Tisk…

Anti-MIP-1-alpha Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5246-100EA 7470 CZK
BIOV5246-100
Anti-MIP-1-alpha Rabbit Polyclonal Antibody
Protilátky
MIP-1α (macrophage inflammatory protein-1α is a 60-amino acid CC chemokine with a molecular weight of 7.9 kDa. MIP-1α induces migration of monocytes/macrophages, B-lymphocytes, activated CD8+ T cells, natural killer (NK) cells, eosinophils. MIP-1α also stimulates basophils to release histamine, induces ICAM-1 expression, mast cell degranulation, and production of TNF-α, IL-1 and IL-6. In addition to its proinflammatory activities, MIP-1α inhibits the proliferation of hematopoietic stem cells in vitro and in vivo.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 4-8 µg/ml

Type: Primary
Antigen: MIP-1-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR