Tisk…

Anti-NT-3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5306-200EA 6910 CZK
BIOV5306-200
Anti-NT-3 Rabbit Polyclonal Antibody
Protilátky
NT-3 is a potent neurotrophic factor that supports the growth and survivability of nerve and/or glial cells. The active form of human NT-3 is a dimer, formed by two identical 119 amino acid subunits, which is held together by strong hydrophobic interactions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: NT-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR