Tisk…

Anti-PANX1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5308-100EA 9170 CZK
BIOV5308-100
Anti-PANX1 Rabbit Polyclonal Antibody
Protilátky
The pannexin gene family encodes a second class of putative gap junction proteins and is highly conserved in invertebrates and mammals. Pannexins (Panx) are four-pass transmembrane proteins that oligomerize to form large pore ion and metabolite-permeable channels. Pannexin-1 (PANX1) and Pannexin-3 are closely related, while Pannexin-2 is a more distant relation. PANX1 is a transmembrane protein that forms a mechanosensitive ATP-permeable channel between adjacent cells and in the endoplasmic reticulum. PANX1 may play a role as a Ca2+ -leak channel to regulate ER Ca2+ homeostasis and regulates neural stem and progenitor cell proliferation.

Recommended Dilutions: Western Blot: 1 - 2?g/ml, ELISA

Type: Primary
Antigen: PANX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR