Tisk…

Anti-RANTES Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5323-100EA 7470 CZK
BIOV5323-100
Anti-RANTES Rabbit Polyclonal Antibody
Protilátky
RANTES (regulated upon activation, normal T-cell expressed and secreted) is a 70-amino acid CC chemokine originally cloned from T-cell-specific sequences-enriched cDNA libraries. Both human and murine RANTES are chemotactic for monocytes. Human RANTES is also a chemoattractant for T-cells and eosinophils, as well as an activator of basophils for histamine release. In contrast to other CC chemokines, RANTES is only slowly expressed after activation of leukocytes. It is involved in a number of pathological conditions like chronic inflammation and autoimmune diseases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: RANTES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR