Tisk…

Anti-ApoSAA Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5324-100EA 6910 CZK
BIOV5324-100
Anti-ApoSAA Rabbit Polyclonal Antibody
Protilátky
Human apo-SAA is a 104 amino acid polypeptide that circulates primarily in association with high-density lipoproteins (HDL;The level of apo-SAA, normally 1-5 µg/ml in plasma, increases 500-1000 fold within 24 hours of an inflammatory stimulus and, under these conditions, is the most abundant HDL apo-lipoprotein. The human SAA gene codes for a 122 amino acid polypeptide, which contains an 18 amino acid N-terminal signal sequence.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ApoSAA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR