Tisk…

Anti-TGF-alpha Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5339-100EA 6910 CZK
BIOV5339-100
Anti-TGF-alpha Rabbit Polyclonal Antibody
Protilátky
Transforming Growth Factor-alpha (TGF-α) stimulates the proliferation of a wide range of epidermal and epithelial cells. TGF-α has been shown to play a role in the autocrine growth of certain transformed cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Neutralization: 2-10 µg/ml

Type: Primary
Antigen: TGF-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR