Tisk…

Anti-TGF-beta2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5343R-100EA 6910 CZK
BIOV5343R-100
Anti-TGF-beta2 Rabbit Polyclonal Antibody
Protilátky
The three mammalian isoforms of TGF-beta (TGF-beta1, TGF-beta2, TGF-beta3) signal throμgh the same receptor and elicit similar biological responses. They are multifunctional cytokines that regulate cell proliferation, growth, differentiation and motility as well as synthesis and deposition of the extracellular matrix. They are involved in various physiological processes including embryogenesis, tissue remodeling and would healing. They are secreted predominantly as latent complexes which are stored at the cell surface and in the extracellular matrix. The release of biologically active TGF-beta isoform from a latent complex involves proteolytic processing of the complex and/or induction of conformational changes by proteins such as thrombospondin-1. TGF-beta2 has been shown to exert suppressive effects on IL-2 dependent T-cell growth, and may also have an autocrine function in enhancing tumor growth by suppressing immuno-surveillance of tumor development.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TGF-beta2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR