Tisk…

Anti-TNF-beta Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5345-100EA 7470 CZK
BIOV5345-100
Anti-TNF-beta Rabbit Polyclonal Antibody
Protilátky
Tumor Necrosis Factor-beta (TNF-β) is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells and certain other target cells. Human TNF-β is an 18.6 kDa protein containing 171 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; Neutralization: 2-5 µg/ml

Type: Primary
Antigen: TNF-beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR