Tisk…

Anti-VEGF Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5364R-100EA 6910 CZK
BIOV5364R-100
Anti-VEGF Rabbit Polyclonal Antibody
Protilátky
VEGF (Vascular Endothelial Growth Factor) is a secreted homodimeric protein, secreted by a variety of vascularized tissues. VEGF stimulates endothelial cell growth, angiogenesis, and capillary permeability.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR