Tisk…

Anti-BD-2 Goat Polyclonal Antibody

Dodavatel: BioVision
BIOV5372-100EA 7470 CZK
BIOV5372-100
Anti-BD-2 Goat Polyclonal Antibody
Protilátky
Produced from sera of goats pre-immunized with highly pure (>98%) recombinant human BD-2. The antibody was purified by affinity chromatography employing immobilized human BD-2 matrix.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml

Type: Primary
Antigen: BD-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR