Tisk…

Anti-GFP Mouse Monoclonal Antibody [clone: 1]

Dodavatel: BioVision
BIOV5410-200EA 7470 CZK
BIOV5410-200
Anti-GFP Mouse Monoclonal Antibody [clone: 1]
Protilátky
EGFP is an improved variant of GFP (green fluorescent protein) with intense green fluorescence. It is an ideal tool for monitoring the dynamics of tagged proteins for protein-protein integration, cell-sorting, organelle labeling, protein localization, gene expression and more. The anti-eGFP antibody recognizes over expressed recombinant proteins containing the eGFP tag fused to either the amino- or carboxy termini of targeted proteins in transfected mammalian cells or other expression systems.

Recommended Dilutions: Western Blot: 0.5-10 µg/ml;

Type: Primary
Antigen: GFP
Clonality: Monoclonal
Clone: 1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: All Species
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR