Tisk…

Anti-mCherry Mouse Monoclonal Antibody [clone: 1]

Dodavatel: BioVision
BIOV5411-100EA 8740 CZK
BIOV5411-100
Anti-mCherry Mouse Monoclonal Antibody [clone: 1]
Protilátky
mCherry is an engineered derivative of one of a family of proteins originally isolated from Cnidarians (jelly fish, sea anemones and corals;mCherry is the second generation monomeric red fluorescent protein that have improved brightness and photostabilty. It is an ideal tool for monitoring the dynamics of tagged proteins for protein-protein integration, cell-sorting, organelle labeling, protein localization, gene expression and more. The anti-mCherry antibody recognizes over expressed recombinant proteins containing the mCherry tag fused to either the amino- or carboxy termini of targeted proteins in transfected mammalian cells or other expression systems.

Recommended Dilutions: Western Blot: 10 µg/ml;

Type: Primary
Antigen: mCherry
Clonality: Monoclonal
Clone: 1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All Species
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR