Tisk…

Anti-BAFF Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5453-100EA 7780 CZK
BIOV5453-100
Anti-BAFF Rabbit Polyclonal Antibody
Rat BAFF (for B cell activating factor belonging to the TNF family) is a newly discovered novel ligand of the TNF family. BAFF plays an important role as co-stimulator of B cell proliferation and function. Recombinant rat BAFF is a soluble protein containing 184 amino acid residues (MW 20.7 kDa;

Recommended Dilutions: Western Blot: at 1-4 µg/ml

Type: Primary
Antigen: BAFF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR