Tisk…

Anti-Midkine Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5479-100EA 6910 CZK
BIOV5479-100
Anti-Midkine Rabbit Polyclonal Antibody
Protilátky
Human Midkine (MK) is a new member of the heparin-binding neurotrophic factor family. MK and PTN (Pleiotrophin) are structural homologs, and are highly conserved among species. MK plays important roles in development and carcinogenesis and has several important biological effects, including promotion of neurite extension and neuronal survival. Human midkine is a 13.4 kDa protein, comprising of 123 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-1 µ/ml; ELISA: 1-4 µg/ml

Type: Primary
Antigen: Midkine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR